Learn More
AdipoGen Progranulin (human) ELISA Kit (Twin Plex),ELISA
SDP
Supplier:  AdipoGen AG45A0018YTPKI01
Encompass_Preferred
Progranulin (human) ELISA Kit (Twin Plex), AdipoGen Life Sciences. ELISA Assay. Detects human progranulin. Colorimetric assay. Sample Types: Cell Culture Supernatant, Plasma, Serum, Urine. Range: 0.063 to 4ng/ml. Sensitivity: 32pg/ml. Progranulin (PGRN) is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration (FTLD). In addition, PGRN has also been linked to tumorigenesis. New cutting-edge publications have shown that progranulin might be a biomarker not only for FTLD, but also for other types of Alzheimer¿s disease (AD). This new data even supposes the potential of progranulin to be a biomarker for MCI (Mild Cognitive Impairment), which would be a hallmark in AD research.
Catalog No. 50-101-5786
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.