Learn More
AdipoGen Progranulin (human) (rec.) (untagged),Protein
SDP
Supplier:  AdipoGen AG40A0188YC050
Encompass_Preferred
Progranulin (human) (rec.) (untagged), AdipoGen Life Sciences. Protein. Signal peptide and human progranulin (aa 1-593) is untagged. Reflects the native sequence with no additional aa. Source: HEK 293 cells. Endotoxin content: <0.01EU/¿g purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >98% (SDS-PAGE). Progranulin (PGRN) is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration (FTLD). In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer¿s Disease (AD) and potentially for MCI (Mild Cognitive Impairment).
Catalog No. 50-463-983
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.