Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proguard™ Professional Handcream
SCI_ED

Catalog No. S66872 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S66872 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S66872 Supplier Decon Laboratories, Inc. Supplier No. 9516/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protects from prolonged glove use and frequent hand scrubbing.

  • Unscented handcream protects from chapping and irritation
  • Particularly effective for improving latex glove comfort
  • Nonpetroleum, nongreasy and absorbs instantly
  • Latex-safe

Order Info

Convenient 16 oz. pump.

Specifications

Product Type Professional Hand Cream
Color White
Form Cream
For Use With (Application) Protects from prolonged glove use and frequent hand scrubbing.
Packaging Pump
Quantity 1 Ea.
Volume (English) 16 oz.
Volume (Metric) 473.1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.