Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech PROK1 Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P188G100
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P188G100 100 μg
500P188G50 50 μg
500P188G1MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P188G100 Supplier PeproTech Supplier No. 500P188G100UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: AVITGACERD VQCGAGTCCA ISLWLRGLRM CTPLGREGEE CHPGSHKVPF FRKRKHHTCP CLPNLLCSRF PDGRYRCSMD LKNINF. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human EG-VEGF. Anti-Human EG-VEGF-specific antibody was purified by affinity chromatography employing an immobilized Human EG-VEGF matrix. Sandwich ELISA: To detect Human EG-VEGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human EG-VEGF (500-P188GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human EG-VEGF. Western Blot: To detect Human EG-VEGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human EG-VEGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PROK1
Gene Accession No. P58294
Gene Alias black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186
Gene Symbols PROK1
Host Species Goat
Immunogen E.coli-derived Recombinant Human EG-VEGF.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84432
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.