Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prokineticin Receptor 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575371 Supplier Invitrogen™ Supplier No. PA575371
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

GPR73A is a receptor for prokineticin 1. It is a member of the GPCR family and is coupled to the G(q) subclass of heteromeric G proteins. Activation of the receptor leads to mobilization of calcium, stimulation of phosphoinositide turnover and activation of p44/p42 mitogen-activated protein kinase. GPR73A expression has been reported in adrenal, peripheral blood leukocytes, brain, colon, pancreas, prostate, rectum, salivary gland, small intestine, spleen, stomach, testis, and thyroid. No human ESTs have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prokineticin Receptor 1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02mg sodium azide; pH 7.2
Gene PROKR1
Gene Accession No. Q8R416, Q8TCW9, Q9JKL1
Gene Alias EG-VEGF receptor; EG-VEGFR1; G protein-coupled receptor 73; G protein-coupled receptor ZAQ; GPCR73A; GPR73; GPR73a; G-protein coupled receptor 73; G-protein coupled receptor ZAQ; PKR1; PK-R1; Prokineticin receptor 1; Prokr1; ZAQ
Gene Symbols PROKR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 20-70 of Human PROKR1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10887, 192648, 58182
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.