Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ ProKlenz Booster Detergent

Catalog No. 17183980 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
200 gal.
300 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-183-980 200 gal.
17-183-979 300 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-183-980 Supplier STERIS™ Supplier No. 1609D3
Add to Cart
Add to Cart
Request a Quote

ProKlenz Booster High Performance Detergent Additive is used for boosting the performance of both alkaline and acidic cleaners and for manual cleaning applications. Its surfactancy and oxidizing chemistry allow for a multitude of cleaning mechanisms. pH - undiluted 5.5, typical

ProKlenz Booster High Performance Detergent Additive can resolve your most challenging process cleaning challenges by greatly enhancing the performance of other detergents with a state-of-the-art surfactant system and oxidizing chemistry.

HOW PROKLENZ BOOSTER WORKS

ProKlenz Booster Detergent Additive is a proprietary blend of surfactants, an oxidizing agent and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.

WHY PROKLENZ BOOSTER?
  • Productivity - Enhances cleaning performance of other detergents or works individually with its superior blend of surfactants and oxidizing chemistry. An excellent product for tablet coating removal in pharmaceutical and dietary supplement manufacturing facilities.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics (above 50°C) when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Globally Availability - phosphate free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Physical Properties: pH - undiluted 5.5, typical

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 5.5 (Undiluted)
Scent Unscented
Sterility Non-sterile
Container Type Tote
Size 200 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.