Learn More
STERIS ProKlenz Foam High Performance Alkaline Cleaner
Catalog No. 17010355 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
30 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-355 30 gal.
17-010-356 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-355 Supplier STERIS Supplier No. 143102WR
Add to Cart
Add to Cart
ProKlenz Foam High Performance Alkaline Cleaner is a globally available all-purpose cleaner for spray applications. With a high foaming system, this product decreases overall labor costs by increasing contact time.
  • ProKlenz Foam High Performance Alkaline Cleaner is an engineered all-purpose cleaner to accommodate the pharmaceutical, biotechnology, cosmetic, and nutraceutical industries worldwide. ProKlenz FOAM Cleaner has a high foam profile, even at a low concentration, for improved cleaning over existing foam products. The free rinsing formula and surfactant system minimize utility consumption and promote cost-effective cleaning processes.
  • When used as directed with suitable foaming spray equipment, ProKlenz FOAM Cleaner can be applied on internal and external processing equipment constructed from common types of metals and plastics. Emulsification and dispersion properties create an effective foam profile for cleaning fats, oils, and organic soils.
  • Globally available product enables harmonized cleaning protocols across sites
  • Free rinsing formula minimizes utility consumption and detergent residue
  • Alkaline compatible surfactant system cleans efficiently with one high foaming product
  • A high foaming system and low use-concentration creates a robust foam profile that promotes cost-effective cleaning and minimizes cleaning time
  • Manufactured within an FDA registered plant, ISO 9001:2015 and ISO 13485:2016 compliant, to meet the highest standard in manufacturing and quality assurance
Specifications
Product Type Detergent
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Size 30 gal.