Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Prolactin ELISA Kit for Rats

Catalog No. MP07DE9966 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
96 Wells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07DE9966 96 Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07DE9966 Supplier MP Biomedical™ Supplier No. 07DE9966

Quantitative Enzyme-linked Immunosorbent Assay

The Prolactin ELISA Kit for Rats is intended for the quantitative determination of prolactin concentration in rat samples.
TRUSTED_SUSTAINABILITY

Specifications

Description Intended for the quantitative determination of prolactin concentration in rat samples.
Product Type Kit and Assay
Sample Type Serum
Target Species Rat
For Use With (Application) Animal Research
Quantity 96 Wells
Regulatory Status For research use only (RUO)
Sample Volume 25 μL
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.