Learn More
Description
MA1-10597 detects Prolactin from human, mouse, rat, bovine, ovine and squirrel monkey samples. MA1-10597 has been successfully used in ELISA, immunohistochemistry, and Western blot applications. The MA1-10597 immunogen is sheep and human Prolactin.
Specifications
Specifications
| Antigen | Prolactin |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 6F11 |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PRL |
| Gene Accession No. | P01236, P01237, P01239, P01240, P06879 |
| Gene Alias | AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
