Learn More
Description
MIP0201 targets Prolactin in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MIP0201 immunogen is human Prolactin. MIP0201 detects Prolactin which has a predicted molecular weight of approximately 25 kDa. MIP0201 was formerly sold as a Seradyn product.
Specifications
Specifications
| Antigen | Prolactin |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | P4C4A12 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PRL |
| Gene Accession No. | P01236 |
| Gene Alias | AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
