Learn More
ILEX LIFE SCIENCES LLC Multus Biotechnology Proliferum LSR 10X Concentrated Serum Substitute, 100 ml (1000 ml diluted)
SDP
Supplier:  ILEX LIFE SCIENCES LLC PLSR001100ML
Encompass_Preferred
Proliferum LSR is a serum-free 1:1 FBS replacement suitable for primary and immortalized cell proliferation. Multus Biotechnology has optimized its performance to work in a variety of environments including adherent culture, suspension culture, cell isolation, cryopreservation and recovery while maintaining cell proliferation and functionality over multiple passages. Fetal Bovine Serum (FBS) is often used in cell culture but is unethically sourced and has large price and performance fluctuations. Serum-free formulations are typically expensive, do not always perform well across the different cell lines, and are not designed for scale. Proliferum LSR serum substitute is designed to grow different species' mammalian myoblasts, fibroblasts and adipocytes. It has the benefits of being serum-free, adaptation-free and demonstrates high performance and versatility across cell types and scaffolding materials over multiple passages.
Catalog No. 50-233-9493
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.