Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Promega 1000 Barrier Tips, 768/pk, 100-1,000microl

Catalog No. PRA1563 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Volume (Metric):
100 to 1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
PRA1563 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA1563 Supplier Promega Supplier No. A1563
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Promega Barrier Tips provide a self-sealing barrier with the convenience of sample retrieval. In retention tests, Promega Barrier Tips virtually eliminated tip retention and sample holdup.

  • Presterilized and certified RNase− and DNase−free
  • Supplied packaged and sealed in covered trays
  • Designed to fit perfectly on all major brands of pipettor

Specifications

Volume (Metric) 100 to 1000 μL
Volume (Metric) Dispensing 100 to 1000 μL
Filtered Filtered
Sterility Sterile
Clean Claims RNase-/DNase-free
For Use With (Application) Radioactive Isotopes, Tissue Culture Fluids, Infectious Samples and Serological Specimens
Format Hinged Rack Sterile
Tip Style Barrier
Packaging Covered Tray
Quantity 768/pk.
Product Type Barrier Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line Promega
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.