Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ABL1 Kinase Enzyme System

Catalog No. PRV1901 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ABL1 Kinase Enzyme System
ADP-Glo Kinase Assay and ABL1 Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV1901 10 μg ABL1 Kinase Enzyme System
PRV9051 1/Ea. ADP-Glo Kinase Assay and ABL1 Kinase Enzyme System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1901 Supplier Promega Supplier No. V1901
Add to Cart
Add to Cart

Recombinant human ABL1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase implicated in processes of cell differentiation, cell division and others.

  • Easily Screen and Profile ABL1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type ABL1 Kinase Enzyme System
Molecular Weight (g/mol) 135
Content And Storage <-65°C
For Use With (Application) Cell differentiation, Cell division, Cell adhesion and stress response
Quantity 10 μg
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.