Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ABL1 Kinase Enzyme System

Catalog No. PRV9051 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ABL1 Kinase Enzyme System
ADP-Glo Kinase Assay and ABL1 Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV9051 1/Ea. ADP-Glo Kinase Assay and ABL1 Kinase Enzyme System
PRV1901 10 μg ABL1 Kinase Enzyme System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9051 Supplier Promega Supplier No. V9051
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ABL1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase implicated in processes of cell differentiation, cell division and others.

  • Easily Screen and Profile ABL1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type ADP-Glo Kinase Assay and ABL1 Kinase Enzyme System
Includes 1 x 10μg ABL1 Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 135
Content And Storage Components at different temps
For Use With (Application) Both primary screening as well as Kinase selectivity profiling
Quantity 1/Ea.
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.