Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega AKT1 Kinase Enzyme System

Catalog No. PRV9061 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9061 1/Ea.
PRV1911 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9061 Supplier Promega Supplier No. V9061
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

AKT1 Kinase Enzyme System (V1911)

  • Kinase: AKT1, 10μg (Human, recombinant; amino acids 695–end). MW: ∽85kDa
  • Substrate: Akt (PKB) substrate (CKRPRAASFAE); derived from the N-terminus of GSK3.
  • Reaction Buffer, DTT, MnCl
ADP-Glo™ Kinase Assay and AKT1 Kinase Enzyme System (V1911)
  • 1 x 10μg AKT1 Kinase Enzyme System (V1911) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant full-length human AKT1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. AKT1/PKB alpha is a serine/threonine kinase that belongs to the AKT family.

  • Easily Screen and Profile AKT1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg AKT1 Kinase Enzyme system (V1911) and 1 x 1000 ADP-Glo™ Kinase assays (V9101), ADP-Glo™ Kinase assay
Content And Storage Components at different temps
For Use With (Application) For both primary screening as well as Kinase selectivity profiling, measures ADP formed from a Kinase reaction
Quantity 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.