Learn More
Promega AMP-Glo™ Assay
Catalog No. PRV5012 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
1000 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV5012 10,000 Assays
PRV5011 1000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV5012 Supplier Promega Supplier No. V5012
Add to Cart
Add to Cart
Request a Quote
The AMP-Glo™ Assay is a homogeneous assay that generates a luminescent signal from any biochemical reaction that produces AMP as a reaction product. This versatile system can measure the activity of a broad range of enzymes.
  • Monitor AMP Concentration in Biochemical Reactions
  • Use with virtually with any AMP-producing enzyme
  • Well-suited for low-activity enzymes
  • Sensitive to low AMP concentration
Specifications
Content And Storage -30°C to -10°C
Detectable Analytes cyclic AMP-specific Phosphodiesterases, Aminoacyl-tRNA synthetases, DNA ligases, Ubiquitin ligases
Type AMP-Glo™ Assay
Includes AMP 10mM, Ultra Pure ATP 10mM, Kinase-Glo™ One Solution 10mL, AMP-Glo™ R2, AMP-Glo™ reagent I
Quantity 10,000 Assays