Learn More
Promega Antibiotic G-418 Sulfate
Catalog No. PRV7983 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
5g
Catalog No.
PRV7983 5g
1 options
Catalog No. PRV7983 Supplier Promega Supplier No. V7983
An aminoglycosidic antibiotic toxic to both prokaryotic and eukaryotic cells.
  • Available as a powder (5g) or as a liquid (20ml)
  • Liquid formulation requires no sterilization