Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega AttoPhos™ AP Fluorescent Substrate System

Catalog No. PRS1000 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Substrate Quantity:
1 x 36mg
3 x 36mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Substrate Quantity
PR-S1000 3 x 36mg
PR-S1001 1 x 36mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-S1000 Supplier Promega Supplier No. S1000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

AttoPhos Substrate, AttoPhos Buffer, AttoPhos Calibration Solution (Mfr. No. S1000 only)

A highly sensitive, flluorescent alkaline phosphatase substrate.

  • Sensitive Fluorescent Alkaline Phosphatase (AP) Substrate
  • Low fluorescence signal until enzymatically activated
  • Detection of AP to 0.1 attomole
  • Linear kinetics over five orders of magnitude of AP concentration

Specifications

Form Powder
Substrate Type AP (Alkaline Phosphatase) Substrate
Includes 3 x 36mg AttoPhos substrate, 4 x 60mL AttoPhos buffer, 1 x 30mL AttoPhos calibration solution
Storage Requirements 4°C
Substrate Quantity 3 x 36mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.