Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega AXL Kinase Enzyme System

Catalog No. PRV9171 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
1000 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9171 1000 Assays
PRV3961 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9171 Supplier Promega Supplier No. V9171
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human AXL (amino acids 473-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. AXL is a member of the receptor tyrosine kinase family, which has oncogenic potential and is implicated in human myeloid leukemia.

  • Easily Screen and Profile AXL Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg AXL Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 55
Content And Storage -70°C
For Use With (Application) Ideal for both primary screening as well as Kinase selectivity profiling
Quantity 1000 Assays
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.