Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Calpain-Glo™ Assay System

Catalog No. PRG8501 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8501 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG8501 Supplier Promega Supplier No. G8501
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Calpain-Glo™ Protease Assay is a homogeneous, luminescent assay that measures calpain 1 (μ) and 2 (m) activities

  • Homogeneous, Luminescent Assay that Measures Calpain 1 (micro) and 2 (m) Activities
  • Simple add-mix-measure protocol
  • Maximum sensitivity while enzyme is still active
  • 1,000x more sensitive than fluorometric assays

  • U.S. Pat. No. 6,602,677, Australian Pat. No. 754312 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.

Specifications

Includes 1 x 10mL Calpain-Glo™ Buffer, 1 x 100μL Suc-LLVY-Glo™ substrate, 1 x 1 Luciferin Detection Reagent
Content And Storage -30°C to -10°C
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.