Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega CAMK4 Kinase Enzyme System

Catalog No. PRV2951 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo Kinase Assay and CAMK4 Kinase Enzyme System
CAMK4 Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV2951 10 μg CAMK4 Kinase Enzyme System
PRV9091 1/Ea. ADP-Glo Kinase Assay and CAMK4 Kinase Enzyme System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV2951 Supplier Promega Supplier No. V2951
Only null left
Add to Cart
Add to Cart

Recombinant full-length human CAMK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK4 is a multifunctional serine/threonine protein kinase and a member of Ca2+/calmodulin-dependent protein kinase family.

  • Easily Screen and Profile CAMK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type CAMK4 Kinase Enzyme System
Content And Storage <-65°C
For Use With (Application) Involved in neural activity-dependent signaling in neuronal nucleus
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.