Learn More
Promega Carboxy Flexi™ Cloning System and Vectors
Catalog No. PRC9401 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
pF5A CMV-neo vector
Catalog No.
PRC9401 pF5A CMV-neo vector
1 options
Catalog No. PRC9401 Supplier Promega Supplier No. C9401
The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences.
  • Untagged Proteins Expressed in Mammalian Cells with a Selectable Marker
  • Expresses high-levels of protein from cloned insert with CMV promoter
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or cell-free protein expression) as needed
  • Adapts to high-throughput formats for large screening projects, enhancing productivity
Order Info
Kit supplied with all components necesssary for 50 transfer reactions; individual vectors sold separately