Learn More
Promega CDK1/CyclinA2 Kinase Enzyme System
Catalog No. PRV2961 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV2961 10 μg
1 options
Catalog No. PRV2961 Supplier Promega Supplier No. V2961
Includes
  • Kinase: CDK1/CyclinA2, 10μg (Human, recombinant full length). MW: ∽59kDa (CDK1); 78kDa (CyclinA2)
  • Substrate: Histone H1- Native histone H1 was purified from calf thymus tissues
  • Others: Reaction Buffer, DTT
Recombinant full-length human CDK1 and CyclinA2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. CDK1 is essential for completion of START, controlling event in the cell cycle required to initiate mitosis.
  • Easily Screen and Profile CDK1/CyclinA2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg