Learn More
Promega CDK2/CyclinA2 Kinase Enzyme System
Catalog No. PRV2971 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV2971 10 μg
1 options
Catalog No. PRV2971 Supplier Promega Supplier No. V2971
Includes
  • Kinase: CDK2/CyclinA2, 10μg (Human, recombinant full-length). M.W.: ∽58kDa (CDK2); ∽78kDa (CyclinA2)
  • Substrate: Native histone H1 was purified from calf thymus tissues
  • Others: Reaction Buffer, DTT
Recombinant full-length human CDK2 and CyclinA2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. CDK2 is a member of the Cyclin-Dependent Kinase family that is ubiquitously expressed.
  • Easily Screen and Profile CDK2/CyclinA2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg