Learn More
Promega CheckMate™/Flexi™ Vector Mammalian Two-Hybrid System
Catalog No. PRC9360 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
PRC9360 Each
1 options
Catalog No. PRC9360 Supplier Promega Supplier No. C9360
Includes
5μg pFN10A (ACT) Flexi Vector, 5μg pFN11A (BIND) Flexi Vector, 20μg pGL4.31[luc2P/GAL4UAS/Hygro] Vector, 20μg pBIND Vector, 20μg pACT Vector, 20μg pBIND-Id Control Vector, 20μg pACT-MyoD Control Vector
The CheckMate/Flexi Vector Mammalian Two-Hybrid System provides a means to study suspected interactions between two proteins or domains and can also be used to generate stable cell lines for cell-based assays.
  • Study Mammalian Protein Interactions, Expression and Post-Translational Modifications
  • Confirm, validate and study suspected protein or domain interactions
  • Generate stable cell lines for cell-based assays
  • Enables mammalian protein expression and post-translational modification studies in situ
Specifications
Content And Storage -30°C to -10°C
Quantity Each
Vector Expression
For Use With (Application) Directional cloning and transfer of Protein coding regions between vectors, can allow Protein expression and post-translational modifications in an environment mimicking the native cell milieu