Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Coelenterazines

Catalog No. PRS2011 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Molecular Weight (g/mol):
407.5
423.5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Molecular Weight (g/mol)
PRS2011 407.5
PRS2001 423.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRS2011 Supplier Promega Supplier No. S2011
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

We offer Coelenterazine (native) and Coelenterazine-h. Coelenterazine (native) is the luminescent substrate for Renilla luciferase and apoaequorin.

  • Supplied as a dried substrate for easy preparation in methanol or ethanol

Specifications

Molecular Weight (g/mol) 407.5
Content And Storage -30°C to -10°C
For Use With (Application) For studying cellular phenomena in expression assays in eukaryotic cells
Quantity 1 x 250 μg
Form Film
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.