Learn More
Promega Collection Plates
Catalog No. PRA9161 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
No. per Pack:
4
Catalog No. No. per Pack
PRA9161 4
1 options
This item is not returnable. View return policy
Catalog No. PRA9161 Supplier Promega Supplier No. A9161
This item is not returnable. View return policy
Collection Plates are designed to use with the Wizard(R) MagneSil(R) Plasmid Purification System for rapid high-throughput isolation of plasmid DNA.
  • Use with the Wizard(R) MagneSil(R) Plasmid Purification System
  • Pack of four 96-well plates
Specifications
Content And Storage 15°C to 30°C
Array 8 x 12
No. of Wells 96
Well Shape Round Bottom (U-well)
No. per Pack 4
Quantity Each