Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Anti-Human IgG (H+L), AP, Secondary Antibody, Promega

Catalog No. PRS3821 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-S3821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-S3821 Supplier Promega Corporation Supplier No. S3821
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for use in protein detection in Western Blots and ELISAs.

  • Ready-to-Use Formulation: No need to dissolve the antibody
  • Applications:
  • Western blotting
  • Enzyme-linked immunosorbent assay (ELISA)
  • Dot blotting

Specifications

Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Description Anti-Human IgG (H+L)
Host Species Goat
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Content And Storage Store Anti-Mouse, Anti-Human, Anti-Rabbit (H+L) (Fc), AP conjugate antibodies at 2° to 10°C. Store Donkey Anti-goat IgG, AP at -30° to -10°C.
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.