Learn More
Promega rLys-C, Mass Spec Grade
Catalog No. PRV1671 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
15 μg
Catalog No. Quantity
PRV1671 15 μg
1 options
Catalog No. PRV1671 Supplier Promega Supplier No. V1671
rLys-C, Mass Spec Grade, is a recombinant Lys-C expressed in E. coli. Sequence origin of rLys-C is Protease IV from Pseudomonas aeruginosa. Similar to a native Lys-C, rLys-C cleaves at the carboxyl side of lysine residues with exceptional specificity.
  • Recombinant Lys-C Cleaves at C-Terminus of Lysines
  • Optimal activity in the pH range of 8-9
  • Retains activity in denaturing conditions such as 8M urea
  • Available lyophilized or frozen
Specifications
Quantity 15 μg