Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Deoxyuridine Triphosphate (dUTP)

Catalog No. PRU1335 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
dATP, dCTP, dGTP, and dUTP
dUTP
Quantity:
10 μM
40 μM
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PRU1335 dUTP 10 μM
PR-U1245 dATP, dCTP, dGTP, and dUTP 40 μM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRU1335 Supplier Promega Supplier No. U1335
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

dUTP (2'-Deoxyuridine, 5'-Triphosphate) can be used in place of dTTP in PCR and RT-PCR to prevent carryover from previous amplifications.

  • High-Quality dNTPs, Including dUTP
  • Highly pure (≥99% triphosphate)
  • Each dNTP supplied at 100mM in water for ease-of-use
  • Incorporate dUTP into PCR products and treat with uracil-N-glycosylase to inhibit subsequent amplifications

Specifications

pH 7.5
Product Type dUTP
Content And Storage -30°C to -10°C
Concentration 100 mM
Purity >99%
Quantity 10 μM
For Use With (Application) cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.