Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega EGFR Kinase Enzyme System

Catalog No. PRV9261 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9261 1/Ea.
PRV3831 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9261 Supplier Promega Supplier No. V9261
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

EGFR Kinase Enzyme System (V3831)

  • Kinase: EGFR, 10μg (Human, recombinant; amino acids 695–end). MW: ∽89kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9261)
  • 1 x 10μg EGFR Kinase Enzyme System (V3831) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human EGFR (amino acids 695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EGFR is the receptor for members of the EGF family and is a transmembrane glycoprotein that has tyrosine kinase activity.

  • Easily Screen and Profile EGFR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg EGFR Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 89
Content And Storage Components at different temps
Quantity 1/Ea.
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.