Learn More
Promega EPHA1 Kinase Enzyme System
Catalog No. PRV3561 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3561 10 μg
PRV9271 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3561 Supplier Promega Supplier No. V3561
Only null left
Add to Cart
Add to Cart
Recombinant mouse EPHA1 (amino acids 569-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EPHA1 is a member of the Eph family of receptor tyrosine kinases that have been implicated in mediating developmental events.
  • Easily Screen and Profile EPHA1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage Store the Kinase at -70°C and the rest of the components at -20°C
Quantity 10 μg