Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FAK Kinase Enzyme System

Catalog No. PRV9301 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9301 1/Ea.
PRV1971 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9301 Supplier Promega Supplier No. V9301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 1 x 10μg CSK Kinase Enzyme System (V1971) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human FAK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. FAK is involved in signal transduction from integrin-enriched focal adhesion sites that mediate cell contact with the extracellular matrix.

  • Easily Screen and Profile FAK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg FAK Kinase Enzyme system, 1 x 1000 assays ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 35
Content And Storage Components at different temps
Quantity 1/Ea.
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.