Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FLT1 Kinase Enzyme System

Catalog No. PRV3001 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3001 10 μg
PRV9331 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3001 Supplier Promega Supplier No. V3001
Add to Cart
Add to Cart

Recombinant human FLT1 (amino acids 784-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FLT1, also known as fms-like tyrosine kinase 1, is a member of the VEGFR family and is related to the oncogene ROS.

  • Easily Screen and Profile FLT1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 94
Content And Storage <-65°C
Quantity 10 μg
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.