Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega β-Galactosidase Enzyme Assay System with Reporter Lysis Buffer

Catalog No. PRE2000 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-E2000 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-E2000 Supplier Promega Supplier No. E2000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

β-galactosidase, 100U; Assay 2X Buffer, 10mL; Reporter Lysis 5X Buffer, 30mL (also available separately, Mfr. No. E3971); Sodium carbonate, 35mL

The Beta-Galactosidase Enzyme Assay System with Reporter Lysis Buffer is used to assay beta-galactosidase activity in cell lysates.

  • Colorimetric Detection of β-Gal Activity
  • Safe, non-isotopic assay
  • Can be used in 96-well plate format
  • Reporter Lysis Buffer allows luciferase, CAT and β-gal assays to be performed from the same cell extract

†Certain applications of this product may require licenses from others.

Specifications

Concentration 2 X
pH 11.6
Content And Storage -30°C to -10°C
For Use With (Application) For assaying beta-galactosidase activity in Lysates prepared from cells transfected with beta-galactosidase reporter vectors
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.