Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Glo Lysis Buffer (GLB)

Catalog No. PRE2661 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE2661 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE2661 Supplier Promega Supplier No. E2661
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A proprietary formulation developed to promote rapid lysis (5 minutes) of cultured mammalian cells without scraping or freeze-thaw cycles.

  • Promotes Rapid Lysis of Cultured Mammalian Cells
  • No need for cell scraping or freeze-thaw cycles
  • Cells lyse within 5 minutes
  • Compatible with Bright-Glo™, Steady-Glo(R), ONE-Glo™ and Renilla-Glo(R) Luciferase Assay Reagents

Specifications

Concentration 1 X
Content And Storage -30°C to +10°C
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.