Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega DB-15 Communication Cable for GloMax™-Multi+ Detection System with Instinct™ Software

Catalog No. PRE8081 Shop All Promega Corporation Products
Change view
Click to view available options
Type:
DB-15 Communication Cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
PRE8081 DB-15 Communication Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE8081 Supplier Promega Supplier No. E8081

For use with GloMax-Multi+ Detection System with Instinct Software.

  • For Use with the GloMax(R)-Multi and Multi+ Detection Systems
  • GloMax(R)-Mulit and Multi+ Microplate Multimode Reader instruments are discontinued
  • Accessories, modules and instrument service for GloMax(R)-Multi available until June 1, 2018
  • Accessories, modules and instrument service for GloMax(R)-Multi+ available until August 1, 2020

Specifications

Type DB-15 Communication Cable
For Use With (Equipment) GloMax-Multi+ Detection System with Instinct Software
Includes 1/ea
Storage Requirements 15 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.