Learn More
Promega GloMax™-Multi Jr Single-Tube Multimode Reader: Optical Kits
Shop All Promega Corporation ProductsDescription
- For Use With the GloMax(R)-Multi Jr Single-Tube Multimode Reader
- Instrument is discontinued
- Accessories (pending availability) and Service Agreement available until November 1, 2020
Specifications
Specifications
| Color | Green |
| For Use With (Equipment) | Single-Tube Multimode Reader |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.