Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GloMax™-Multi Jr Single-Tube Multimode Reader: Optical Kits

Catalog No. p-4903288 Shop All Promega Corporation Products
Change view
Click to view available options
Color:
Blue
GFPUV
Green
Red
UV
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
PRE6073 Green
PRE6071 Blue
PRE6072 UV
PRE6074 Red
PRE6075 GFPUV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PRE6073 Supplier Promega Supplier No. E6073

Fluorescence Kits for use with the GloMax-Multi Jr Single-Tube Multimode Reader. The GloMax-Multi Jr is designed to provide the utmost flexibility. In addition to high performance, the GloMax-Multi Jr blends user-friendly operation and a small footprint with flexible purchasing options.

  • For Use With the GloMax(R)-Multi Jr Single-Tube Multimode Reader
  • Instrument is discontinued
  • Accessories (pending availability) and Service Agreement available until November 1, 2020

Specifications

Color Green
For Use With (Equipment) Single-Tube Multimode Reader
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.