Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Glycosidases

Catalog No. PRV4871 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10,000 U
50,000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV4871 10,000 U
PRV4875 50,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV4871 Supplier Promega Supplier No. V4871
Add to Cart
Add to Cart

Endo H is cloned from S. plicatus and overexpressed in E. coli.

  • Characterize Protein Glycosylation
  • Monitor protein trafficking
  • Characterize glycan location and structure
  • Does not cleave complex glycans
  • Supplied at a concentration of 500u/μL

Specifications

Concentration 500 U/μL
For Use With (Application) Determining the state of Protein Glycosylation, characterizing Glycan structure
Formulation 1 x 10000U Endo H, 1 x 1mL 10X Endo H Reaction Buffer, 1 x 1mL 10X Denaturing Solution
Content And Storage -30°C to -10°C
Quantity 10,000 U
Product Type Endoglycosidase H
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.