Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Fetuin

Catalog No. PRV4961 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV4961 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV4961 Supplier Promega Supplier No. V4961
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fetuin is a glycoprotein with O-linked and N-linked glycosylation sites.

  • Glycoprotein with O- and N-Linked Glycosylation Sites
  • Deglycosylation substrate control for PNGase F, EndoH activity monitoring
  • Supplied at a concentration of 10mg/ml

Specifications

Concentration 10 mg/mL
For Use With (Application) Determining the state of Protein Glycosylation, characterizing Glycan structure
Formulation 1 x 50μL Fetuin
Content And Storage -30°C to -10°C
Quantity 500 μg
Product Type Fetuin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.