Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTEV™ Protease

Catalog No. PRG6601 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG6601 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG6601 Supplier Promega Supplier No. G6601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HaloTEV Protease (81kDa) is a fusion between the HaloTag™ protein and TEV protease, a highly specific proteolytic enzyme that cleaves at a specific TEV site, a specific seven-amino-acid sequence (ENLYFQ(G/S)).

  • Cleave and Recover Target Protein in a Single Step
  • Easy to covalently remove HaloTEV from your purified protein with HaloLink™ Resin
  • Proteins can be purified tag-free in a single step as the HaloLink™ Resin will bind both HaloTag(R) protein tag and the HaloTEV protease
  • Supplied at a concentration of 5u/μL

Specifications

Product Type HaloTEV Protease
Quantity 1000 U
Content And Storage <−65°C
For Use With (Application) Covalent immobilization on HaloLink™ Resin and removal from a cleavage reaction in a single-step purification, improves purity of the final target Protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.