Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Herring Sperm DNA

Catalog No. PRD1816 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-D1816 500 mg
PR-D1811 10 mg
PR-D1815 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-D1816 Supplier Promega Supplier No. D1816
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

Produces fragments with a size range of 80% between 100-3,000 base pairs; free of DNase or RNase activity.

  • Quality tested and certified free of DNase or RNase activity
  • Use as a blocking agent in hybridizations or as carrier DNA
  • Provided as a ready-to-use 10mg/mL solution

Specifications

Content And Storage -30°C to -10°C
Concentration 10 μg/μL
DNase Yes
For Use With (Application) Blocking agent in hybridizations or as carrier DNA
Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.