Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
- Cells are ready to use—no preparation time necessary
- Transformation efficiencies guaranteed
- No worries about undesirable recombination events (recA- mutation), and no nuclease carryover in miniprep DNA (endA- mutation)
Specifications
Specifications
| Concentration | >108cfu/μg |
| Content And Storage | Store at -70°C. |
| For Use With (Application) | Transformation of unmethylated DNA derived from PCR, cDNA and many other sources |
| Includes | Single-Use JM109 Competent Cells |
| pH | 7 |
| Product Type | Competent Cells |
| Quantity | 20 x 0.05 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?