Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Kanamycin Positive Control RNA

Catalog No. PRC1381 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-C1381 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-C1381 Supplier Promega Supplier No. C1381
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1.2kb Kanamycin Positive Control RNA is supplied in 10mM Tris-HCl (pH 7.4), 1mM EDTA.

Specifications

Content And Storage <-65°C
For Use With (Application) Synthesis of double stranded cDNA from poly(A)+ RNA
Product Type 1.2kb Kanamycin Positive Control RNA
Quantity 5 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.