Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega KDR Kinase Enzyme System

Catalog No. PRV9471 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV9471 1/Ea.
PRV2681 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9471 Supplier Promega Supplier No. V9471
Add to Cart
Add to Cart

Includes

KDR Kinase Enzyme System (V2681)

  • Kinase: KDR, 10μg (Human, recombinant; amino acids 789–end), M.W.: ∽110kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT
ADP-Glo™ Kinase Assay and KDR Kinase Enzyme System (V9471)
  • 1 x 10μg KDR Kinase Enzyme System (V2681) and 1 x 1000 ADP-Glo™ Kinase Assays (V9471)

Recombinant human KDR (amino acids 789-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. KDR (or kinase insert domain receptor) is a growth factor receptor tyrosine kinase originally isolated from human endothelial cells.

  • Easily Screen and Profile KDR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 1 x 10μg KDR Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 110
Content And Storage Components at different temps
Quantity 1/Ea.
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.