Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Kinase-Glo™ Max Luminescent Kinase Assay System

Catalog No. PRV6074 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6074 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV6074 Supplier Promega Supplier No. V6074
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.

  • Monitors Activity of Purified Kinases by Quantifying ATP
  • Luminescent signal is inversely proportional to the amount of kinase activity
  • Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP

Specifications

Content And Storage -30°C to -10°C
Quantity 10 x 100 mL
Form Fluid

The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.