Learn More
Description
Includes
6X Blue/Orange Loading Dye
- Six DNA Fragments of 3,530-21,226bp
- Load 1μL/lane
- Supplied at a concentration of 0.5μg/μL
- Supplied with 6X Blue/Orange Loading Dye
Specifications
Specifications
| Buffer | 10mM Tris HCl pH 7.8 |
| Color | Blue, Orange |
| Concentration | 0.5 μg/μL |
| Form | Liquid |
| Formulation | 100μg lambda DNA/EcoRI marker, 1mL blue/orange 6X loading dye |
| For Use With (Application) | Separation, size determination and quantitation of DNA and RNA fragments |
| No. of Bands | 6 |
| Quantity | 100 μg |
| Storage Requirements | −30 to −10°C |
| Product Type | DNA Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.