Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega LigaFast™ Rapid DNA Ligation System

Catalog No. PRC6711 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
DNA Ligation System
Ligation Buffer
Quantity:
1.5 mL
150 Reactions
30 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PR-C6711 Ligation Buffer 1.5 mL
PR-M8221 DNA Ligation System 30 Reactions
PR-M8225 DNA Ligation System 150 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PR-C6711 Supplier Promega Supplier No. C6711
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed to eliminate the need for further purification prior to transformation of ligated DNA

  • Use with 5', 3' or blunt-ended DNA inserts
  • Ligation of cohesive ends in 5 minutes, blunt ends in 15 minutes at room temperature
  • No requirement to purify ligated DNA prior to heat-shock transformation in E. coli ligations conducted at room temperature
  • No additional buffer modifications required prior to use
  • Ligations performed using the LigaFast™ system are comparable to standard overnight ligations
  • Blue/White Cloning Qualified

Specifications

Content And Storage -30°C to -10°C
Formulation Tris HCl (pH 7) 10mM, KCl 50mM, DTT 1mM, EDTA 0.1mM, Glycerol 50%
For Use With (Application) 5′, 3′ or blunt ended DNA inserts
pH 7.8
Product Type Ligation Buffer
Quantity 1.5 mL
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.