Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Luciferase SP6/T7/T3 Control DNAs

Catalog No. PRL4821 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-L4821 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-L4821 Supplier Promega Supplier No. L4821
Only null left
Add to Cart
Add to Cart

Luciferase SP6 and T7 Control DNAs are used with the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter.

  • Functional Controls for the TnT(R) Quick Coupled and TnT(R) Coupled Transcription/Translation Systems
  • Contains luciferase gene under transcriptional control of an SP6 or T7 phage RNA polymerase promoter
  • Includes a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene
  • Easy to monitor control reactions with a luciferase assay

1The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.

Specifications

Product Type Luciferase T7 Control DNA
Storage Buffer TE Buffer
Content And Storage -30°C to -10°C
Concentration 0.5 mg/mL
Quantity 20 μg
For Use With (Application) TnT quick coupled and TnT toupled transcription/translation systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.